ENA296275417 Enamine

2,4-dichloro-6-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD09733327
  • Cas Number : 568586-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40106
Availability details for
Quantity
  • Availability
  • Add to Cart