ENA296275426 Enamine

cyclohexyl(cyclopropyl)methanone

  • MDL number MFCD12153378
  • Cas Number : 58688-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65682
Availability details for
Quantity
  • Availability
  • Add to Cart