ENA296275428 Enamine

1-(3-methoxypropyl)piperidin-4-amine

  • MDL number MFCD11104531
  • Cas Number : 179474-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65687
Availability details for
Quantity
  • Availability
  • Add to Cart