ENA296275435 Enamine

2-nitropyridin-3-yl 4-methylbenzene-1-sulfonate

  • MDL number MFCD16622173
  • Cas Number : 1240527-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65697

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65697
Availability details for
Quantity
  • Availability
  • Add to Cart