ENA296275437 Enamine

5-chloro-3-(2-chlorophenyl)-1,2,4-oxadiazole

  • MDL number MFCD12774193
  • Cas Number : 1178490-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64483

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-64483
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart