ENA296275442 Enamine

2-chloro-5-isothiocyanatopyridine

  • MDL number MFCD16622177
  • Cas Number : 323588-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65705

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P285,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P310,P311,P312,P330,P342+P311,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-65705
pictograms GHS05,GHS06,GHS08
hstatements H302,H312,H314,H331,H334
Availability details for
Quantity
  • Availability
  • Add to Cart