ENA296275443 Enamine

tert-butyl N-(cyanomethyl)-N-ethylcarbamate

  • MDL number MFCD09953055
  • Cas Number : 1240529-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65706
Availability details for
Quantity
  • Availability
  • Add to Cart