ENA296275445 Enamine

3-(pyrrolidin-1-yl)pyrrolidine-2,5-dione

  • MDL number MFCD16622178
  • Cas Number : 7685-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123...125
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65708

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65708
Availability details for
Quantity
  • Availability
  • Add to Cart