ENA296275449 Enamine

1-methyl-1H-indole-4-carboxylic acid

  • MDL number MFCD08435874
  • Cas Number : 90924-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65727
Availability details for
Quantity
  • Availability
  • Add to Cart