ENA296275451 Enamine

4-methyl-N-(2-phenylethyl)-1,3-thiazol-2-amine

  • MDL number MFCD07696128
  • Cas Number : 196796-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65730
Availability details for
Quantity
  • Availability
  • Add to Cart