ENA296275452 Enamine

[5-(4-chlorophenyl)-1,3,4-oxadiazol-2-yl]methanol

  • MDL number MFCD01755369
  • Cas Number : 54014-07-2
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 126-128
Iupac Name [5-(4-chlorophenyl)-1,3,4-oxadiazol-2-yl]methanol
Inchi Code 1S/C9H7ClN2O2/c10-7-3-1-6(2-4-7)9-12-11-8(5-13)14-9/h1-4,13H,5H2
Inchi Key MJZPGYRFLJFAKG-UHFFFAOYSA-N

Description

Melting Point 126-128
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65731

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-65731
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart