ENA296275457 Enamine

2-(cyclopentyloxy)-2-phenylacetic acid

  • MDL number MFCD12145314
  • Cas Number : 39773-76-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65736
Availability details for
Quantity
  • Availability
  • Add to Cart