ENA296275464 Enamine

1-(4-aminophenyl)-N-(prop-2-en-1-yl)methanesulfonamide

  • MDL number MFCD12791555
  • Cas Number : 88919-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65744
Availability details for
Quantity
  • Availability
  • Add to Cart