ENA296275469 Enamine

1-cyclohexylcyclopropan-1-ol

  • MDL number MFCD16622184
  • Cas Number : 112650-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-65753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65753
Availability details for
Quantity
  • Availability
  • Add to Cart