ENA296275472 Enamine

5-fluoro-2-(piperidin-1-yl)aniline

  • MDL number MFCD04971023
  • Cas Number : 252758-87-5
  • Molecular Weight : 194.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65756
Availability details for
Quantity
  • Availability
  • Add to Cart