ENA296275477 Enamine

2-chloro-N-methyl-N-(1,3-thiazol-2-yl)acetamide

  • MDL number MFCD01924772
  • Cas Number : 878657-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46878
Availability details for
Quantity
  • Availability
  • Add to Cart