ENA296275480 Enamine

3-[(cyanomethyl)sulfamoyl]-4-methylbenzoic acid

  • MDL number MFCD13197835
  • Cas Number : 1051333-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65773
Availability details for
Quantity
  • Availability
  • Add to Cart