ENA296275485 Enamine

7-hydroxy-2,2-dimethyl-2,3-dihydro-1-benzofuran-6-carboxylic acid

  • MDL number MFCD11203619
  • Cas Number : 1038374-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65794
Availability details for
Quantity
  • Availability
  • Add to Cart