ENA296275486 Enamine

2-chloro-N-(cyclopropylmethyl)-N-methylacetamide

  • MDL number MFCD12798934
  • Cas Number : 1178287-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48394
Availability details for
Quantity
  • Availability
  • Add to Cart