ENA296275487 Enamine

3-(methoxymethyl)-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD11205090
  • Cas Number : 1094424-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65798
Availability details for
Quantity
  • Availability
  • Add to Cart