ENA296275489 Enamine

2-(piperidin-3-yl)acetic acid hydrochloride

  • MDL number MFCD02181158
  • Cas Number : 71985-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65801
Availability details for
Quantity
  • Availability
  • Add to Cart