ENA296275493 Enamine

3-[(2-methoxy-2-oxoethyl)sulfanyl]propanoic acid

  • MDL number MFCD11646610
  • Cas Number : 93274-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65811
Availability details for
Quantity
  • Availability
  • Add to Cart