ENA296275494 Enamine

[(4-fluorophenyl)methyl][2-(pyridin-2-yl)ethyl]amine

  • MDL number MFCD11143778
  • Cas Number : 344304-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65812
Availability details for
Quantity
  • Availability
  • Add to Cart