ENA296275499 Enamine

2-(chloromethyl)-5-(propan-2-yl)-1,3,4-oxadiazole

  • MDL number MFCD08059899
  • Cas Number : 3914-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65822
Availability details for
Quantity
  • Availability
  • Add to Cart