ENA296275504 Enamine

2-(3-methylbutan-2-yl)-2,3-dihydro-1H-isoindol-5-amine

  • MDL number MFCD11655022
  • Cas Number : 1099612-87-9
  • Molecular Weight : 204.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64657
Availability details for
Quantity
  • Availability
  • Add to Cart