ENA296275510 Enamine

N-[(4-methoxyphenyl)methyl]-2-methyl-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD12598214
  • Cas Number : 1157739-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65854
Availability details for
Quantity
  • Availability
  • Add to Cart