ENA296275516 Enamine

2-[2-(propan-2-yl)-1,3-thiazol-4-yl]ethan-1-amine

  • MDL number MFCD09971285
  • Cas Number : 202817-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64697

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64697
Availability details for
Quantity
  • Availability
  • Add to Cart