ENA296275519 Enamine

N-(4-hydroxyphenyl)-4-methylbenzamide

  • MDL number MFCD00442533
  • Cas Number : 23646-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65872
Availability details for
Quantity
  • Availability
  • Add to Cart