ENA296275521 Enamine

1-(1H-indazole-5-carbonyl)piperidine-4-carboxylic acid

  • MDL number MFCD13667975
  • Cas Number : 1099083-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65878
Availability details for
Quantity
  • Availability
  • Add to Cart