ENA296275523 Enamine

methyl 3-(4-formyl-3,5-dimethyl-1H-pyrazol-1-yl)propanoate

  • MDL number MFCD12405235
  • Cas Number : 949020-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65880
Availability details for
Quantity
  • Availability
  • Add to Cart