ENA296275536 Enamine

4-[(2-aminopyridin-3-yl)oxy]butanenitrile

  • MDL number MFCD10694090
  • Cas Number : 1152561-49-3
  • Molecular Weight : 177.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-65930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65930
Availability details for
Quantity
  • Availability
  • Add to Cart