ENA296275537 Enamine

3-(butan-2-yl)-1-(2,6-diethylphenyl)thiourea

  • MDL number MFCD12634921
  • Cas Number : 66608-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-65931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65931
Availability details for
Quantity
  • Availability
  • Add to Cart