ENA296275540 Enamine

2,4-dioxo-1,3-diazaspiro[4.5]decane-8-carboxylic acid

  • MDL number MFCD16622203
  • Cas Number : 21129-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283-285
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65934
Availability details for
Quantity
  • Availability
  • Add to Cart