ENA296275545 Enamine

(2-cyanoethyl)(dimethylsulfamoyl)methylamine

  • MDL number MFCD05695648
  • Cas Number : 850802-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65942
Availability details for
Quantity
  • Availability
  • Add to Cart