ENA296275549 Enamine

1,3-dichloropropan-2-yl chloroformate

  • MDL number MFCD16622204
  • Cas Number : 55183-48-7
  • Molecular Weight : 191.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65949
Availability details for
Quantity
  • Availability
  • Add to Cart