ENA296275558 Enamine

1-benzylcyclopentan-1-ol

  • MDL number MFCD01106286
  • Cas Number : 2015-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-60
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65985
Availability details for
Quantity
  • Availability
  • Add to Cart