ENA296275559 Enamine

2-amino-3-phenyl-1-(piperidin-1-yl)propan-1-one hydrochloride

  • MDL number MFCD09028973
  • Cas Number : 879006-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64715
Availability details for
Quantity
  • Availability
  • Add to Cart