ENA296275562 Enamine

5-chloro-6-[(5-fluoro-2-methylphenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD12617927
  • Cas Number : 1040006-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65995
Availability details for
Quantity
  • Availability
  • Add to Cart