ENA296275568 Enamine

1-(pyridin-4-yl)-1H-pyrazol-4-amine

  • MDL number MFCD01692456
  • Cas Number : 28466-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-159
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66023
Availability details for
Quantity
  • Availability
  • Add to Cart