ENA296275587 Enamine

2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD12066280
  • Cas Number : 1014630-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64736
Availability details for
Quantity
  • Availability
  • Add to Cart