ENA296275593 Enamine

2-(pentan-2-yl)-2,3-dihydro-1H-isoindol-5-amine

  • MDL number MFCD11655050
  • Cas Number : 1094463-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64753
Availability details for
Quantity
  • Availability
  • Add to Cart