ENA296275601 Enamine

tert-butyl N-(2-aminoethyl)-N-ethylcarbamate

  • MDL number MFCD04114276
  • Cas Number : 105628-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64766

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-64766
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart