ENA300790634 Enamine

methyl 2-cyclopropyl-4-hydroxy-6-methylpyrimidine-5-carboxylate

  • MDL number MFCD09863299
  • Cas Number : 950032-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35003

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35003
Availability details for
Quantity
  • Availability
  • Add to Cart