ENA300790638 Enamine

3-(1-methyl-1H-pyrazol-5-yl)piperidine

  • MDL number MFCD16990615
  • Cas Number : 1251925-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50599

Safety Information

msds https://www.enaminestore.com/catalog/EN300-50599
Availability details for
Quantity
  • Availability
  • Add to Cart