ENA300790651 Enamine

tert-butyl N-[1-(N-hydroxycarbamimidoyl)cyclobutyl]carbamate

  • MDL number MFCD16817402
  • Cas Number : 1251924-20-5
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 1-[(hydroxyamino)(imino)methyl]cyclobutylcarbamate
Inchi Code 1S/C10H19N3O3/c1-9(2,3)16-8(14)12-10(5-4-6-10)7(11)13-15/h15H,4-6H2,1-3H3,(H2,11,13)(H,12,14)
Inchi Key NJCTYVNAPFQCMS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-63220

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-63220
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart