ENA300790653 Enamine

tert-butyl N-(1-carbamothioylcyclobutyl)carbamate

  • MDL number MFCD16817404
  • Cas Number : 1251924-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63223
Availability details for
Quantity
  • Availability
  • Add to Cart