ENA300790657 Enamine

sodium 2-[4-(4-methanesulfonylphenyl)-1,3-thiazol-2-yl]acetate

  • MDL number MFCD16817408
  • Cas Number : 1251925-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65089
Availability details for
Quantity
  • Availability
  • Add to Cart