ENA300790658 Enamine

2-amino-5-ethyl-3H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one

  • MDL number MFCD16817409
  • Cas Number : 1251924-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65166
Availability details for
Quantity
  • Availability
  • Add to Cart