ENA300790659 Enamine

6-chloro-3-(piperidin-4-yl)-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD16817410
  • Cas Number : 1251924-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-65337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65337
Availability details for
Quantity
  • Availability
  • Add to Cart