ENA300790660 Enamine

5-(5-chloro-2-fluorophenyl)-1H-pyrazol-3-amine

  • MDL number MFCD16817411
  • Cas Number : 1136906-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65506
Availability details for
Quantity
  • Availability
  • Add to Cart